Learn More
IKA™ RV 10.84 Evaporation Flask
Product Code. 11845922 Shop All IKA Products
Change view
Click to view available options
Volume (Metric):
1000 mL
Unit Size:
Each
Product Code. Volume (Metric) Unit Size
11845922 1000 mL Each
1 options
Product Code. 11845922 Supplier IKA™ Supplier No. 0003740500
  • ONLINE EXCLUSIVE PRICE
    104.60 €
    93.50 € / Pack of 1
    Save 11.10 €
    10% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-quality borosilicate glass flask designed for evaporation processes.
  • High-quality borosilicate glass construction
  • Suitable for evaporation processes
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass
Type Evaporating Flask
Standard Taper Joint Number Center NS 29/32
Volume (Metric) 1000 mL