missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate, 98+%
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Description
Used as a chiral Bronsted acid catalyst in the enantoselective Mannich reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as a chiral Bronsted acid catalyst in the enantoselective Mannich reaction.
Solubility
Soluble in DMSO and methanol (hot).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
Used as a chiral Bronsted acid catalyst in the enantoselective Mannich reaction.
Solubility
Soluble in DMSO and methanol (hot).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
Specifications
Specifications
| Melting Point | >300°C |
| Quantity | 1 g |
| Beilstein | 4787775 |
| Solubility Information | Soluble in DMSO and methanol (hot). |
| Optical Rotation | +605° (c=1 in Methanol) |
| Formula Weight | 348.3 |
| Percent Purity | ≥98% |
| Chemical Name or Material | (S)-(+)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction