missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate, 98+%

Product Code. 15410837
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15410837

Brand: Thermo Scientific Alfa Aesar L14137.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 35193-64-7 | C20H13O4P | 348.294 g/mol

Used as a chiral Bronsted acid catalyst in the enantoselective Mannich reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a chiral Bronsted acid catalyst in the enantoselective Mannich reaction.

Solubility
Soluble in DMSO and methanol (hot).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35193-64-7
Molecular Formula C20H13O4P
Molecular Weight (g/mol) 348.294
MDL Number MFCD00010045
InChI Key JEHUZVBIUCAMRZ-UHFFFAOYSA-N
Synonym 1,1'-binaphthyl-2,2'-diyl hydrogenphosphate, s-+-1,1'-binaphthyl-2,2'-diyl hydrogenphosphate, r---1,1'-binaphthyl-2,2'-diyl hydrogenphosphate, 1,1'-binaphthyl-2,2'-diyl hydrogen phosphate, r---bnp acid, 11bs-4-hydroxydinaphtho 2,1-d:1',2'-f 1,3,2 dioxaphosphepine 4-oxide, r---1,1'-binaphthyl-2,2'-diyl hydrogen phosphate, r-4-hydroxydinaphtho 2,1-d:1',2'-f 1,3,2 dioxaphosphepine 4-oxide, s-+-bnp acid, 4-hydroxydinaphtho 2,1-d:1',2'-f 1,3,2 dioxaphosphepine 4-oxide
PubChem CID 99589
SMILES C1=CC=C2C(=C1)C=CC3=C2C4=C(C=CC5=CC=CC=C54)OP(=O)(O3)O

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 4787775
Solubility Information Soluble in DMSO and methanol (hot).
Optical Rotation +605° (c=1 in Methanol)
Formula Weight 348.3
Percent Purity ≥98%
Chemical Name or Material (S)-(+)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.