missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1-(1-Naphthyl)ethyl isocyanate, 95%

Product Code. 11329096
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11329096 250 mg 250mg
11319096 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11329096 Supplier Thermo Scientific Alfa Aesar Supplier No. L20242.MD
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 73671-79-1 | C13H11NO | 197.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73671-79-1
Molecular Formula C13H11NO
Molecular Weight (g/mol) 197.24
MDL Number MFCD00064181
InChI Key GONOHGQPZFXJOJ-UHFFFAOYNA-N
Synonym s-+-1-1-naphthyl ethyl isocyanate, 1-1s-1-isocyanatoethyl naphthalene, s-nei, s-1-1-naphthyl ethyl isocyanate, isocyanic acid s-+-1-1-naphthyl ethyl ester, 1-1-isocyanatoethyl naphthalene #, pubchem8101, s-+-1-naphthyl ethyl isocyanate, s--1-1-naphthyl ethyl isocyanate, s-+-1-1-naphthyl ethylisocyanate
PubChem CID 7023510
IUPAC Name 1-[(1S)-1-isocyanatoethyl]naphthalene
SMILES CC(N=C=O)C1=C2C=CC=CC2=CC=C1

Specifications

Density 1.118
Boiling Point 106°C to 108°C (0.16 mmHg)
Flash Point 93°C (199°F)
Refractive Index 1.6045
Quantity 250 mg
UN Number UN2206
Beilstein 4675069
Sensitivity Moisture sensitive; May form a precipitate
Optical Rotation +47° (c=3.5 in Toluene)
Formula Weight 197.24
Percent Purity 95%
Chemical Name or Material (S)-(+)-1-(1-Naphthyl)ethyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.