missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-(1-Naphthyl)ethylamine, 99%

Product Code. 11423397
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11423397 1 g 1g
11433397 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11423397 Supplier Thermo Scientific Alfa Aesar Supplier No. A17176.03
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10420-89-0 | C12H14N | 172.25 g/mol

(S)-(-)-1-(1-Naphthyl)ethylamine, (S)-(-)-1-(1-Naphthyl)ethylamine is used in the asymmetric synthesis of α-cyanocarboxylates. Also used in the synthesis of chiral imadazolin-2-ylidene ligands used in organometallic catalysis. Chiral/Asymmetric synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-1-(1-Naphthyl)ethylamine, (S)-(-)-1-(1-Naphthyl)ethylamine is used in the asymmetric synthesis of α-cyanocarboxylates. Also used in the synthesis of chiral imadazolin-2-ylidene ligands used in organometallic catalysis. Chiral/Asymmetric synthesis.

Solubility
Soluble in chloroform, ethanol.

Notes
Air Sensitive. Stable under recommended storage conditions. Keep away from acids, acid chlorides, acid anhydrides, oxidizing agents, chloroformates.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10420-89-0
Molecular Formula C12H14N
Molecular Weight (g/mol) 172.25
MDL Number MFCD00064179
InChI Key RTCUCQWIICFPOD-VIFPVBQESA-O
Synonym s---1-1-naphthyl ethylamine, s-1-naphthalen-1-yl ethanamine, 1s-1-naphthalen-1-yl ethan-1-amine, s---alpha-1-naphthyl ethylamine, s-1-1-naphthyl ethylamine, 1s-1-1-naphthyl ethanamine, s---1-naphthyl ethylamine, s-alpha-methyl-1-naphthalenemethanamine, 1s-1-naphthalen-1-yl ethanamine
PubChem CID 66325
IUPAC Name (1S)-1-naphthalen-1-ylethanamine
SMILES C[C@H]([NH3+])C1=C2C=CC=CC2=CC=C1

Specifications

Density 1.06
Boiling Point 152°C to 153°C (11 mmHg)
Flash Point >110°C (230°F)
Odor Amine-like
Refractive Index 1.623
Quantity 1 g
UN Number UN2922
Beilstein 2208024
Sensitivity Air Sensitive
Solubility Information Soluble in chloroform,ethanol.
Formula Weight 171.24
Percent Purity 99%
Chemical Name or Material (S)-(-)-1-(1-Naphthyl)ethylamine
Show More Show Less
compliance-icons
Product Identifier
  • (S)-(-)-1-(1-Naphthyl)ethylamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: (S)-(-)-1-(1-Naphthyl)ethylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.