missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-(2-Naphthyl)ethylamine, ChiPros 99+%, ee 99+%

Product Code. 11393916
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11393916 1 g 1g
11303926 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11393916 Supplier Thermo Scientific Alfa Aesar Supplier No. L17024.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3082-62-0 | C12H13N | 171.243 g/mol

(S)-(-)-1-(2-Naphthyl)ethylamine is used in asymmetric synthesis. It is also used in the preparation of thiourea compounds using chiral amine, which is used as a bifunctional catalyst for the Strecker and nitro-Michael reactions. Further, it acts as a chiral building block and chiral resolution reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-1-(2-Naphthyl)ethylamine is used in asymmetric synthesis. It is also used in the preparation of thiourea compounds using chiral amine, which is used as a bifunctional catalyst for the Strecker and nitro-Michael reactions. Further, it acts as a chiral building block and chiral resolution reagent.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Sensitive to carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3082-62-0
Molecular Formula C12H13N
Molecular Weight (g/mol) 171.243
MDL Number MFCD00085366
InChI Key KHSYYLCXQKCYQX-VIFPVBQESA-N
Synonym s-1-naphthalen-2-yl ethanamine, s---1-2-naphthyl ethylamine, s-1-2-naphthyl ethylamine, 1s-1-naphthalen-2-yl ethan-1-amine, 2-naphthalenemethanamine, alpha-methyl-, alphas, 1s-1-2-naphthyl ethylamine, 1s-1-naphthalen-2-yl ethanamine, s-1-2-naphthyl ethanamine, s-1-naphth-2-yl ethylamine, s-1-naphth-2-yl-ethylamine
PubChem CID 1201503
IUPAC Name (1S)-1-naphthalen-2-ylethanamine
SMILES CC(C1=CC2=CC=CC=C2C=C1)N

Specifications

Melting Point 48°C to 52°C
Boiling Point 90°C (0.15 mmHg)
Quantity 1 g
UN Number UN3259
Beilstein 3195949
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Optical Rotation −20° (c=1 in Methanol)
Formula Weight 171.24
Percent Purity ≥99%
Assay ee ≥99%
Chemical Name or Material (S)-(-)-1-(2-Naphthyl)ethylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.