missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-(3-Chlorophenyl)ethylamine, ChiPros™, 98%, ee 98+%, Thermo Scientific™

Product Code. 11496590
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496590 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11496590 Supplier Thermo Scientific Alfa Aesar Supplier No. H27637.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
As active pharmaceutical drugs and intermediates possess different biological activities.

Notes
Moisture Sensitive. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air. Refrigerated (2-4°C).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68297-62-1
Molecular Formula C8H10ClN
Molecular Weight (g/mol) 155.63
MDL Number MFCD06761823
InChI Key DQEYVZASLGNODG-UHFFFAOYNA-N
Synonym s-1-3-chlorophenyl ethanamine, s-1-3-chlorophenyl ethylamine, 1s-1-3-chlorophenyl ethanamine, 1s-1-3-chlorophenyl ethan-1-amine, s-3-chloro-, a-methylbenzylamine, s---1-3-chlorophenyl ethylamine, s-1-3-chlorophenyl ethylamine, chipros, benzenemethanamine, 3-chloro-, a-methyl-,, as, s-3-chloro-alpha-methylbenzylamine, chipros r , produced by basf
PubChem CID 2507691
IUPAC Name (1S)-1-(3-chlorophenyl)ethanamine
SMILES CC(N)C1=CC(Cl)=CC=C1

Specifications

Quantity 1 g
UN Number UN2735
Boiling Point 112°C (13 mmHg)
Formula Weight 155.63
Percent Purity 98%
Assay ee ≥98%
Chemical Name or Material (S)-1-(3-Chlorophenyl)ethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.