missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-(4-Methylphenyl)ethylamine, ChiPros 98%, ee 99+%

Product Code. 11309105
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11309105 1 g 1g
11319105 5 g 5g
11329105 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11309105 Supplier Thermo Scientific Alfa Aesar Supplier No. L16329.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 27298-98-2 | C9H13N | 135.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27298-98-2
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00145246
InChI Key UZDDXUMOXKDXNE-UHFFFAOYNA-N
Synonym s-1-p-tolyl ethanamine, s---1-p-tolyl ethylamine, s---4-1-aminoethyl toluene, s-1-p-tolyl ethylamine, 1s-1-4-methylphenyl ethan-1-amine, 1s-1-p-tolyl ethanamine, 1s-1-4-methylphenyl ethanamine, s-1-p-tolylethanamine, 1-4-methylphenyl ethanamine #, s-1-4-methylphenyl ethylamine
PubChem CID 7015759
SMILES CC(N)C1=CC=C(C)C=C1

Specifications

Melting Point <-20°C
Density 0.94
Boiling Point 204°C
Flash Point 82°C (179°F)
Refractive Index 1.5215
Quantity 1 g
UN Number UN2735
Beilstein 2422644
Sensitivity Air Sensitive
IUPAC Name 1-(4-methylphenyl)ethan-1-amine
Formula Weight 135.21
Percent Purity 98%
Assay ee ≥99%
Chemical Name or Material (S)-(-)-1-(4-Methylphenyl)ethylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.