missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1-Amino-2-propanol, 98%, Thermo Scientific™

Product Code. 11338975
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11338975 250 mg 250mg
11328975 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11338975 Supplier Thermo Scientific Alfa Aesar Supplier No. L14102.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2799-17-9
Molecular Formula C3H9NO
Molecular Weight (g/mol) 75.11
MDL Number MFCD00064429
InChI Key HXKKHQJGJAFBHI-UHFFFAOYNA-N
Synonym s-+-1-amino-2-propanol, s-1-aminopropan-2-ol, 2s-1-aminopropan-2-ol, +-isopropanolamine, s-1-amino-2-propanol, 2-propanol, 1-amino-, 2s, 1-amino-2-propanol #, 2-propanol, 1-amino-, s, ambotzfal1033
PubChem CID 7311736
IUPAC Name 1-aminopropan-2-ol
SMILES CC(O)CN

Specifications

Melting Point 24°C to 26°C
Density 0.96
Boiling Point 160°C
Flash Point 71°C (159°F)
Refractive Index 1.449
Quantity 250 mg
UN Number UN3259
Beilstein 1718868
Sensitivity Air sensitive; Hygroscopic
Optical Rotation +18.5° (c=1.8 in Water)
Formula Weight 75.11
Percent Purity 98%
Chemical Name or Material (S)-(+)-1-Amino-2-propanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.