missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-BOC-3-hydroxypiperidine, 97%, Thermo Scientific Chemicals

Product Code. 11996361 Shop All Thermo Scientific Chemicals Products
Cambia vista
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Dimensione della confezione:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity Packaging Dimensione della confezione
11996361 1 g Glass bottle 1g
11906371 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 11996361 Fornitore Thermo Scientific Acros N. del fornitore 436440010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 143900-44-1 | Molecular Formula: C10H19NO3 | Molecular Weight: 201.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 143900-44-1
Molecular Formula C10H19NO3
Molecular Weight (g/mol) 201.27
MDL Number MFCD04115307
InChI Key UIJXHKXIOCDSEB-UHFFFAOYNA-N
Synonym s-1-boc-3-hydroxypiperidine, s-tert-butyl 3-hydroxypiperidine-1-carboxylate, s-n-boc-3-hydroxypiperidine, tert-butyl 3s-3-hydroxypiperidine-1-carboxylate, s-1-n-boc-3-hydroxy-piperidine, s-1-n-boc-3-hydroxypiperidine, s-n-boc-3-hydroxy piperidine, s-3-hydroxypiperidine, n-boc protected, s-3-hydroxy-piperidine-1-carboxylic acid tert-butyl ester, 3s-1-tert-butoxycarbonyl-3-hydroxypiperidine
PubChem CID 1514399
SMILES CC(C)(C)OC(=O)N1CCCC(O)C1

Specifica

CAS Min % 96.0
CAS Max % 100.0
Melting Point 48.9°C to 51.7°C
Color Tan to White
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation + 9.3
IUPAC Name tert-butyl 3-hydroxypiperidine-1-carboxylate
Formula Weight 201.26
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material (S)-1-BOC-3-Hydroxypiperidine
Vedi altri risultati Mostra meno risultati
compliance-icons
Identificativo prodotto
  • (S)-1-BOC-3-hydroxypiperidine, 97%
Avvertenza
  • Warning
Categoria di pericolo
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicazioni di pericolo
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Consigli di prudenza
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.