missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-Boc-3-methylpiperazine, 98%

Product Code. 11491810
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491810 1 g 1g
11401820 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11491810 Supplier Thermo Scientific Alfa Aesar Supplier No. H27534.03
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 147081-29-6 | C10H20N2O2 | 200.282 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147081-29-6
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.282
MDL Number MFCD02683204
InChI Key FMLPQHJYUZTHQS-QMMMGPOBSA-N
Synonym s-4-n-boc-2-methylpiperazine, s-4-n-boc-2-methyl piperazine, s-tert-butyl 3-methylpiperazine-1-carboxylate, s-4-boc-2-methylpiperazine, s-1-boc-3-methyl-piperazine, tert-butyl 3s-3-methylpiperazine-1-carboxylate, s-1-boc-3-methylpiperazine, s-4bocmp, 4-boc-2-s-methyl-piperazine, s-1-n-boc-3-methylpiperazine
PubChem CID 7023035
IUPAC Name tert-butyl (3S)-3-methylpiperazine-1-carboxylate
SMILES CC1CN(CCN1)C(=O)OC(C)(C)C

Specifications

Melting Point 38°C to 41°C
Quantity 1 g
Sensitivity Air sensitive
Optical Rotation −16° (c=1 in Dioxane)
Formula Weight 200.28
Percent Purity 98%
Chemical Name or Material (S)-(-)-1-Boc-3-methylpiperazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.