missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1-Bromo-2-methylbutane, 97%, stab. with potassium carbonate

Product Code. 11456140
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11456140 5 g 5g
11466140 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11456140 Supplier Thermo Scientific Alfa Aesar Supplier No. H27078.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 534-00-9 | C5H11Br | 151.05 g/mol

(S)-(+)-1-Bromo-2-methylbutane, is used for the synthesis of chiral nematic liquid crystals and optically active Grignard reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-1-Bromo-2-methylbutane, is used for the synthesis of chiral nematic liquid crystals and optically active Grignard reagents.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents, strong bases. Light sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 534-00-9
Molecular Formula C5H11Br
Molecular Weight (g/mol) 151.05
MDL Number MFCD00078662
InChI Key XKVLZBNEPALHIO-YFKPBYRVSA-N
Synonym s-1-bromo-2-methylbutane, s-+-1-bromo-2-methylbutane, butane, 1-bromo-2-methyl-, 2s, unii-cgs982pbtw, 2s-1-bromo-2-methylbutane, d-amyl bromide, cgs982pbtw, 2s-1-bromo-2-methyl-butane, +-2-methylbutyl bromide, butane, 1-bromo-2-methyl-, s
PubChem CID 5464167
IUPAC Name (2S)-1-bromo-2-methylbutane
SMILES CC[C@H](C)CBr

Specifications

Density 1.223
Boiling Point 121°C to 122°C
Flash Point 22°C (79°F)
Refractive Index 1.4455
Quantity 5 g
UN Number UN1993
Beilstein 1718815
Sensitivity Light sensitive; May form a precipitate
Merck Index 14,601
Solubility Information Insoluble in water.
Optical Rotation +4.5° (Neat)
Formula Weight 151.05
Percent Purity 97%
Chemical Name or Material (S)-(+)-1-Bromo-2-methylbutane, stabilized with potassium carbonate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (S)-(+)-1-Bromo-2-methylbutane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: (S)-(+)-1-Bromo-2-methylbutane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.