missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1-Methyl-3-phenylpropylamine, 98%

Product Code. 10347421 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10347421 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10347421 Supplier Thermo Scientific Acros Supplier No. 347660010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 4187-57-9 | C10H15N | 149.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4187-57-9
CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.9360g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C10H15N
Refractive Index 1.5130 to 1.515
MDL Number MFCD00671665
Quantity 1 g
Specific Gravity 0.936
Specific Rotation Condition +10° (20°C c=neat)
Specific Rotation + 10
Synonym 2s-4-phenylbutan-2-amine, s-+-1-methyl-3-phenylpropylamine, unii-atn027167d, s-4-phenylbutan-2-amine, s-4-phenylbutane-2-amine, 4-phenylbutan-2-amine, s, s-1-methyl-3-phenylpropylamine, s-1-methyl-3-phenyl-propylamine, benzenepropanamine, a-methyl-, as, benzenepropanamine, alpha-methyl-, s
InChI Key WECUIGDEWBNQJJ-VIFPVBQESA-N
SMILES CC(CCC1=CC=CC=C1)N
IUPAC Name (2S)-4-phenylbutan-2-amine
Molecular Weight (g/mol) 149.23
PubChem CID 6994564
Formula Weight 149.23
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material (S)-(+)-1-Methyl-3-phenylpropylamine, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (S)-(+)-1-Methyl-3-phenylpropylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H412-Harmful to aquatic life with long lasting effects.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.