missing translation for 'onlineSavingsMsg'
Learn More

S-(1-Oxido-2-pyridyl)-N,N,N',N'-tetramethylthiuronium hexafluorophosphate, 98+%, Thermo Scientific™

Product Code. p-7036036
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15435265 1 g 1g
15445265 5 g 5g
15455265 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15435265 Supplier Thermo Scientific Alfa Aesar Supplier No. H54277.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 212333-72-7
Molecular Formula C10H16F6N3OPS
Molecular Weight (g/mol) 371.282
MDL Number MFCD05865266
InChI Key UCOGEMMJHLHOAW-UHFFFAOYSA-N
Synonym hott, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuronium hexafluorophosphate, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiouronium hexafluorophosphate, hott n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuronium hexafluorophosphate, ambotzrl-1156, acmc-209fij, ksc911c8f, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuron, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuronhexafluorophosphate, n,n,n,n-tetramethyl-s-1-oxido-2-pyridyl thiouronium hexafluorophosphate
PubChem CID 10571248
IUPAC Name [dimethylamino-(1-oxidopyridin-1-ium-2-yl)sulfanylmethylidene]-dimethylazanium;hexafluorophosphate
SMILES CN(C)C(=[N+](C)C)SC1=CC=CC=[N+]1[O-].F[P-](F)(F)(F)(F)F

Specifications

Melting Point 120°C to 125°C
Quantity 1 g
Sensitivity Moisture sensitive
Formula Weight 371.28
Percent Purity ≥98%
Chemical Name or Material S-(1-Oxido-2-pyridyl)-N,N,N',N'-tetramethylthiuronium hexafluorophosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.