missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline, TRC

Product Code. 30135653
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135653 25 g 25g
30138151 5 g 5g
17900425 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135653 Supplier Toronto Research Chemicals Supplier No. TRCP33693025G
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118864-75-8
Molecular Formula C15 H15 N
Molecular Weight (g/mol) 209.29
Synonym Isoquinoline, 1,2,3,4-tetrahydro-1-phenyl-, (S)-, (+)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline, (-)-(S)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline, (1S)-1,2,3,4-Tetrahydro-1-phenylisoquinoline, (1S)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline, (1S)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline, (S)-(+)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline, (S)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline, Solifenacin Succinate Imp. A (EP)
IUPAC Name (1S)-1-phenyl-1,2,3,4-tetrahydroisoquinoline
SMILES C1Cc2ccccc2[C@@H](N1)c3ccccc3

Specifications

Quantity 25 g
InChI Formula InChI=1S/C15H15N/c1-2-7-13(8-3-1)15-14-9-5-4-6-12(14)10-11-16-15/h1-9,15-16H,10-11H2/t15-/m0/s1
Formula Weight 209.1204
Percent Purity >95%
Chemical Name or Material (S)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.