missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-Phenylbutylamine, ChiPros™, 98%, ee 98+%, Thermo Scientific™

Product Code. 11416090
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11416090 5 g 5g
11406090 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11416090 Supplier Thermo Scientific Alfa Aesar Supplier No. H27048.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3789-60-4
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.237
MDL Number MFCD08064291
InChI Key XHOXKVFLASIOJD-JTQLQIEISA-N
Synonym s-1-phenylbutylamine, s-1-phenylbutan-1-amine, 1s-1-phenylbutan-1-amine, pubchem24143, s-alpha-phenylbutylamine, s-1-phenyl-butylamine, s-1-phenyl-1-butanamine, s-1-phenylbutylamine, chipros, benzenemethanamine, a-propyl-, as, s-1-phenylbutylamine, chipros , ee 98+%
PubChem CID 2449444
IUPAC Name (1S)-1-phenylbutan-1-amine
SMILES CCCC(C1=CC=CC=C1)N

Specifications

Boiling Point 220°C
Quantity 5 g
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 149.24
Percent Purity 98%
Assay ee ≥98%
Chemical Name or Material (S)-1-Phenylbutylamine
Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.