missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-Phenylethanol, 97+%

Product Code. 15493227
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15493227 25 g 25g
11329709 1 g 1g
11339709 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15493227 Supplier Thermo Scientific Alfa Aesar Supplier No. B21188.14
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1445-91-6 | C8H10O | 122.17 g/mol

It finds its application in the synthesis of optically active products. Used for the synthesis of various pharmaceutical compounds. It can also be used as a chiral derivatizing agent for determining the absolute configuration of secondary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the synthesis of optically active products. Used for the synthesis of various pharmaceutical compounds. It can also be used as a chiral derivatizing agent for determining the absolute configuration of secondary alcohols.

Solubility
Soluble in water at 20°C 20g/L. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1445-91-6
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00064264
InChI Key WAPNOHKVXSQRPX-ZETCQYMHSA-N
PubChem CID 443135
ChEBI CHEBI:16346
SMILES C[C@H](O)C1=CC=CC=C1

Specifications

Melting Point 9°C to 11°C
Density 1.012
Boiling Point 97°C to 99°C (20 mmHg)
Flash Point 85°C
Odor Mild
Refractive Index 1.528
Quantity 25 g
UN Number UN2937
Beilstein 2039797
Synonym s-1-phenylethanol, s---1-phenylethanol, 1 s-1-phenylethanol, 1 s-1-phenylethan-1-ol, unii-2 mic4qly2x, 1-phenylethanol, s, 2mic4qly2x, s-1-phenethyl alcohol, s-1-phenyl-1-ethanol, s-1-phenylethan-1-ol
Solubility Information Soluble aq. soln. at 20°C 20 g/L Soluble in alcohol.
Optical Rotation -45°C=5 in. Methanol)
IUPAC Name (1 S)-1-phenylethan-1-ol
Formula Weight 122.17
Percent Purity 99%
Chemical Name or Material (S)-(-)-1-Phenylethanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.