missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-Phenylethylamine, 99+%, produced by BASF AG

Product Code. 10512072 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10512072 1 kg Glass bottle 1kg
10378843 50 g Glass bottle 50g
10028911 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10512072 Supplier Thermo Scientific Acros Supplier No. 371450010
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2627-86-3 | C8H11N | 121.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chiral derivatization reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2627-86-3
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00064406
InChI Key RQEUFEKYXDPUSK-ZETCQYMHSA-N
Synonym s---1-phenylethylamine, s-1-phenylethanamine, 1s-1-phenylethanamine, s---alpha-methylbenzylamine, s-1-phenylethylamine, l--alpha-methylbenzylamine, l-alpha-methylbenzylamine, s-alpha-methylbenzenemethanamine, --alpha-phenethylamine, l---1-phenylethylamine
PubChem CID 75818
ChEBI CHEBI:35321
IUPAC Name (1S)-1-phenylethanamine
SMILES CC(C1=CC=CC=C1)N

Specifications

Melting Point -10.0°C
Color Colorless
Density 0.9400g/mL
Boiling Point 187.0°C to 189.0°C
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH(CH3)NH2
Refractive Index 1.5250 to 1.5270
Quantity 1 kg
Beilstein 12,II,587
Fieser 09,363; 13,185; 17,276; 12,319; 11,411; 05,441; 03,199; 01,838; 02,271; 08,393
Merck Index 13, 6054
Specific Gravity 0.94
Specific Rotation Condition −40.00 (20.00°C neat)
Specific Rotation − 40.00
Solubility Information Solubility in water: 42g/L (20°C). Other solubilities: soluble in most organic solvents,miscible with alcohols and ethers
Formula Weight 121.18
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material (S)-(-)-1-Phenylethylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (S)-(-)-1-Phenylethylamine, produced by BASF AG
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.