missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-Phenylethylamine, ChiPros 99+%, ee 99.5%

Product Code. 11389655
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11389655 25 g 25g
11379655 5 g 5g
11399655 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11389655 Supplier Thermo Scientific Alfa Aesar Supplier No. L19118.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 2627-86-3 | C8H11N | 121.183 g/mol

(S)-(-)-1-Phenylethylamine is utilized in a one-pot, multi-component synthesis of a highly substituted, chiral pyrrole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-1-Phenylethylamine is utilized in a one-pot, multi-component synthesis of a highly substituted, chiral pyrrole.

Solubility
Slightly miscible with water.

Notes
Keep away from sources of ignition. Incompatible with acids, acid chlorides, acid anhydrides and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2627-86-3
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00064406
InChI Key RQEUFEKYXDPUSK-ZETCQYMHSA-N
Synonym s---1-phenylethylamine, s-1-phenylethanamine, 1s-1-phenylethanamine, s---alpha-methylbenzylamine, s-1-phenylethylamine, l--alpha-methylbenzylamine, l-alpha-methylbenzylamine, s-alpha-methylbenzenemethanamine, --alpha-phenethylamine, l---1-phenylethylamine
PubChem CID 75818
ChEBI CHEBI:35321
IUPAC Name (1S)-1-phenylethanamine
SMILES CC(C1=CC=CC=C1)N

Specifications

Melting Point -10°C
Density 0.948
Boiling Point 187°C
Flash Point 71°C (159°F)
Odor Amine-like
Refractive Index 1.526
Quantity 25 g
UN Number UN2735
Beilstein 2204907
Sensitivity Air Sensitive
Merck Index 14,6026
Solubility Information Slightly miscible with water.
Optical Rotation −30° (c=10 in Ethanol)
Formula Weight 121.18
Percent Purity ≥99%
Assay ee 99.5%
Chemical Name or Material (S)-(-)-1-Phenylethylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.