missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2,2',6,6'-Tetramethoxy-4,4'-bis(di(3,5-xylyl)phosphino)-3,3'-bipyridine

Product Code. 11445410
Change view
Click to view available options
Quantity:
0.1 g
0.5 g
Unit Size:
0.1g
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11445410 0.5 g 0.5g
11435410 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11445410 Supplier Thermo Scientific Alfa Aesar Supplier No. 044614.02
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
This item is not returnable. View return policy

Asymmetric hydrogenation | CAS: 443347-10-2 | C46H50N2O4P2 | 756.864 g/mol

(S)-(-)-2,2',6,6'-Tetramethoxy-4,4'-bis(di(3,5-xylyl)phosphino)-3,3'-bipyridine is used in Pharmaceutical and industry applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 443347-10-2
Molecular Formula C46H50N2O4P2
Molecular Weight (g/mol) 756.864
MDL Number MFCD04974235
InChI Key JRTHAKOHBMETRC-UHFFFAOYSA-N
Synonym unii-1a4eo1493y, unii-n2pcw56063, r-4,4'-bis bis 3,5-dimethylphenyl phosphino-2,2',6,6'-tetramethoxy-3,3'-bipyridine, s---2,2',6,6'-tetramethoxy-4,4'-bis di 3,5-xylyl phosphino-3,3'-bipyridine, s-4,4'-bis bis 3,5-dimethylphenyl phosphino-2,2',6,6'-tetramethoxy-3,3'-bipyridine, xyl-p-phos, r, xyl-p-phos, s, r-xyl-p-phos mi, s-xyl-p-phos mi, xyl-p-phos
PubChem CID 11182279
IUPAC Name [3-[4-bis(3,5-dimethylphenyl)phosphanyl-2,6-dimethoxypyridin-3-yl]-2,6-dimethoxypyridin-4-yl]-bis(3,5-dimethylphenyl)phosphane
SMILES CC1=CC(=CC(=C1)P(C2=CC(=CC(=C2)C)C)C3=CC(=NC(=C3C4=C(N=C(C=C4P(C5=CC(=CC(=C5)C)C)C6=CC(=CC(=C6)C)C)OC)OC)OC)OC)C

Specifications

Melting Point 158°C to 162°C
Quantity 0.5 g
Solubility Information Sparingly soluble in water.
Formula Weight 756.86
Physical Form Powder
Chemical Name or Material (S)- (-)-2,2',6,6'-Tetramethoxy-4,4'-bis (di (3,5-xylyl)phosphino)-3,3'-bipyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.