missing translation for 'onlineSavingsMsg'
Learn More

(S)-2,2'-Bis(di-p-tolylphosphino)-4,4',6,6'-tetramethoxybiphenyl, 97+%, Thermo Scientific Chemicals

Product Code. 15426106
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15426106 1 g 1g
15436106 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15426106 Supplier Thermo Scientific Alfa Aesar Supplier No. H60921.03
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1365531-82-3 | Molecular Formula: C44H44O4P2 | Molecular Weight: 698.78 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1365531-82-3
Molecular Formula C44H44O4P2
Molecular Weight (g/mol) 698.78
MDL Number MFCD19443623
InChI Key JXFDRZQHVPTIMH-UHFFFAOYSA-N
Synonym s-2,2'-bis di-p-tolylphosphino-4,4',6,6'-tetramethoxybiphenyl, r-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis di-p-tolylphosphine, s-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis di-p-tolylphosphine
PubChem CID 87110704

Specifications

Quantity 1 g
Sensitivity Air sensitive
Formula Weight 698.77
Percent Purity ≥97%
Chemical Name or Material (S)-2,2'-Bis(di-p-tolylphosphino)-4,4',6,6'-tetramethoxybiphenyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.