missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(thiophen-3-yl)propanoic acid, 97%, BLD Pharm™

Product Code. 30804434
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
25 g
Unit Size:
100mg
1g
250mg
25g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30804434 5 g 5g
30797628 25 g 25g
30791081 100 mg 100mg
30791174 1 g 1g
30802555 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30804434 Supplier BLD Pharm Supplier No. BD14675g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 186320-06-9
Molecular Formula C22H19NO4S
MDL Number MFCD00151916
Quantity 5 g
SMILES O=C(O)[C@@H](NC(OCC1C2=C(C3=C1C=CC=C3)C=CC=C2)=O)CC4=CSC=C4
Molecular Weight (g/mol) 393.46
Percent Purity 97%
Chemical Name or Material (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(thiophen-3-yl)propanoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.