missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Amino-1-phenylethanol, 97%

Product Code. 11378696
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11378696 1 g 1g
11388696 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378696 Supplier Thermo Scientific Alfa Aesar Supplier No. L19404.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 56613-81-1 | C8H11NO | 137.18 g/mol

(S)-(+)-2-Amino-1-phenylethanol is used as an intermediate in organic synthesis. It is involved in the preparation of enantiopure (R)-octopamine, (R)-tembamide and (R)-aegeline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-2-Amino-1-phenylethanol is used as an intermediate in organic synthesis. It is involved in the preparation of enantiopure (R)-octopamine, (R)-tembamide and (R)-aegeline.

Solubility
Soluble in methanol.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56613-81-1
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00239405
InChI Key ULSIYEODSMZIPX-MRVPVSSYSA-N
Synonym s-2-amino-1-phenylethanol, s---2-phenylglycinol, 1s-2-amino-1-phenylethanol, s-2-amino-1-phenylethan-1-ol, 1s-2-amino-1-phenylethan-1-ol, s-+-2-amino-1-phenylethanol, r---alpha-aminomethyl benzyl alcohol, pubchem13836, 2-amino-1 s-phenylethanol
PubChem CID 643217
IUPAC Name (1S)-2-amino-1-phenylethanol
SMILES NC[C@@H](O)C1=CC=CC=C1

Specifications

Melting Point 57°C to 63°C
Boiling Point 160°C (17 mmHg)
Quantity 1 g
UN Number UN3259
Beilstein 2412760
Sensitivity Air Sensitive
Solubility Information Soluble in methanol.
Optical Rotation +42° (c=2 in Ethanol)
Formula Weight 137.18
Percent Purity 97%
Chemical Name or Material (S)-(+)-2-Amino-1-phenylethanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.