missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S-(+)-2-Amino-1-propanol, TRC
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
Specifications
Specifications
| Chemical Name or Material | (2S)-2-Aminopropan-1-ol |
| CAS | 2749-11-3 |
| Molecular Formula | C3 H9 N O |
| Quantity | 25 g |
| Synonym | 1-Propanol, 2-amino-, (2S)-, 1-Propanol, 2-amino-, (S)-, 1-Propanol, 2-amino-, L- (8CI), (2S)-2-Amino-1-propanol, ((S)-2-Hydroxy-1-methylethyl)amine, (+)-2-Amino-1-propanol, (+)-2-Aminopropanol, (+)-Alaninol, (2S)-(+)-2-Aminopropan-1-ol, (2S)-(+)-2-Aminopropan-1-ol, (2S)-2-Aminopropan-1-ol, (S)-2-Amino-1-propanol, (S)-2-Amino-1-propanol, (S)-2-Aminopropanol, (S)-Alaninol, 2-(S)-Amino-propan-1-ol, 2S-Aminopropanol, L-(+)-Alaninol, L-2-Amino-1-propanol, L-2-Aminopropanol, L-Alaninol, S-(+)-2-Amino-1-propanol, S-(+)-Alaninol, [(S)-1-(Hydroxymethyl)ethyl]amine |
| InChI Formula | InChI=1S/C3H9NO/c1-3(4)2-5/h3,5H,2,4H2,1H3/t3-/m0/s1 |
| SMILES | C[C@H](N)CO |
| IUPAC Name | (2S)-2-aminopropan-1-ol |
| Molecular Weight (g/mol) | 75.1097 |
| Formula Weight | 75.0684 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction