missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-Amino-3-(naphthalen-1-yl)propanoic acid, 97%, BLD Pharm™

Product Code. 31108311
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
10 g
25 g
100 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31108311 5 g
31096895 100 g
31106378 10 g
31101178 1 g
31103209 25 g
31098460 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 31108311 Supplier BLD Pharm Supplier No. BD146665g
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 55516-54-6
Molecular Formula C13H13NO2
MDL Number MFCD00066086
Quantity 5 g
SMILES O=C(O)[C@@H](N)CC1=C2C=CC=CC2=CC=C1
Molecular Weight (g/mol) 215.25
Percent Purity 97%
Chemical Name or Material (S)-2-Amino-3-(naphthalen-1-yl)propanoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.