missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Amino-4-phenylbutyric acid ethyl ester hydrochloride, 97%, Thermo Scientific™

Product Code. 10743132 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10743132 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10743132 Supplier Thermo Scientific Acros Supplier No. 365670050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 90891-21-7
CAS Min % 96.0
CAS Max % 100.0
Melting Point 153.0°C to 154.0°C
Color Tan to White
Elemental Analysis 56.76 to 61.48% C, 5.52 to 5.98% N
Loss on Drying 1% max. ()
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Molecular Formula C12H17NO2·HCl
MDL Number MFCD00190691
Quantity 5 g
Specific Rotation 39.7
Synonym ethyl s-2-amino-4-phenylbutanoate hydrochloride, l-homophenylalanine ethyl ester hydrochloride, h-hophe-oet.hcl, ethyl l-homoalaninate hcl, s-ethyl 2-amino-4-phenylbutanoate hydrochloride, ethyl 2s-2-amino-4-phenylbutanoate hydrochloride, pubchem6054, h-hph-oet.hcl, h-hph-oet. hcl, homophenylalanine ethyl ester hydrochloride
InChI Key PTFKZMFFSIYCOV-MERQFXBCSA-N
SMILES CCOC(=O)C(CCC1=CC=CC=C1)N.Cl
IUPAC Name ethyl (2S)-2-amino-4-phenylbutanoate;hydrochloride
Molecular Weight (g/mol) 243.73
PubChem CID 2734606
Formula Weight 243.73
Percent Purity 97%
Physical Form Powder
Chemical Name or Material (S)-(+)-2-Amino-4-phenylbutyric acid ethyl ester hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.