missing translation for 'onlineSavingsMsg'
Learn More

S-(2-Aminoethyl)-l-cysteine, hydrochloride, TRC

Product Code. 30012404
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012404 500 mg 500mg
30008928 250 mg 250mg
30010516 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012404 Supplier Toronto Research Chemicals Supplier No. TRCA608055500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4099-35-8
Molecular Formula C5H13ClN2O2S
Molecular Weight (g/mol) 200.69
Synonym S-(2-Aminoethyl)-L-cysteine Monohydrochloride, NSC 186915;S-(2-Aminoethyl)-L-cysteine Hydrochloride;S-(2-Aminoethyl)-L-cysteine Monohydrochloride;S-(beta-Aminoethyl)-L-cysteine Hydrochloride, Thialysine Hydrochloride
IUPAC Name (2R)-2-amino-3-(2-aminoethylsulfanyl)propanoic acid;hydrochloride
SMILES Cl.NCCSC[C@H](N)C(=O)O

Specifications

InChI Formula InChI=1S/C5H12N2O2S.ClH/c6-1-2-10-3-4(7)5(8)9;/h4H,1-3,6-7H2,(H,8,9);1H/t4-;/m0./s1
Quantity 500 mg
Formula Weight 200.0386
Shipping Condition Room Temperature
Chemical Name or Material S-(2-Aminoethyl)-l-cysteine, hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.