missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Aminohexane, ChiPros 99+%, ee 99+%, Thermo Scientific™

Product Code. 11364206
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11364206 25 g 25g
11344206 1 g 1g
11354206 5 g 5g
3 options
This item is not returnable. View return policy

Product Code. 11364206

Brand: Thermo Scientific Alfa Aesar L19305.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(S)-(+)-2-Aminohexane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in water (17g/L at 20°C)

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70492-67-0
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.19
MDL Number MFCD00671626
InChI Key WGBBUURBHXLGFM-UHFFFAOYNA-N
Synonym s-2-aminohexane, 2s-hexan-2-amine, s-2-hexylamine, s-+-2-aminohexane, s-+-2-aminohexane, chipros, ee 99+%, s-2-hexaneamine, pubchem6750, 2s-2-hexanamine, 2-hexanamine, 2s, +-s-2-aminohexane
PubChem CID 6999846
SMILES CCCCC(C)N

Specifications

Melting Point <-20°C
Density 0.755
Boiling Point 116°C
Flash Point −3°C (26°F)
Quantity 25 g
UN Number UN2733
Beilstein 5237601
Sensitivity Air Sensitive
Solubility Information Soluble in water (17g/L at 20°C)
IUPAC Name hexan-2-amine
Formula Weight 101.19
Percent Purity ≥99%
Assay ee ≥99%
Chemical Name or Material (S)-(+)-2-Aminohexane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.