missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Chloropropionic acid, 98%

Product Code. 11489820
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489820 5 g 5g
11499820 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489820 Supplier Thermo Scientific Alfa Aesar Supplier No. L09427.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 29617-66-1 | C3H4ClO2 | 107.51 g/mol

Through the stereo displacement of (S)-(-)-2-Chloropropionic acid PPAR Agonist can be synthesized via asymmetric hydrogenation of a cinnamic acid derivative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Through the stereo displacement of (S)-(-)-2-Chloropropionic acid PPAR Agonist can be synthesized via asymmetric hydrogenation of a cinnamic acid derivative.

Solubility
Fully miscible in water.

Notes
Store in cool dry condition. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29617-66-1
Molecular Formula C3H4ClO2
Molecular Weight (g/mol) 107.51
MDL Number MFCD00064205
InChI Key GAWAYYRQGQZKCR-REOHCLBHSA-M
Synonym s---2-chloropropionic acid, s-2-chloropropanoic acid, s-2-chloropropionic acid, 2s-2-chloropropanoic acid, l-2-chloropropanoic acid, l-2-chloropropionic acid, alpha-l-chloropropionic acid, unii-pcc7j322cm, 2s-chloropropanoic acid, 2 s-chloropropionic acid
PubChem CID 107915
ChEBI CHEBI:73956
IUPAC Name (2S)-2-chloropropanoic acid
SMILES C[C@H](Cl)C([O-])=O

Specifications

Melting Point 4°C
Density 1.27
Boiling Point 187°C
Flash Point 107°C (225°F)
Refractive Index 1.4345
Quantity 5 g
UN Number UN2511
Beilstein 1720257
Solubility Information Fully miscible in water.
Formula Weight 108.53
Percent Purity 99%
Chemical Name or Material (S)-(-)-2-Chloropropionic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.