missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-((Diphenylphosphino)methyl)pyrrolidine, 97%, Thermo Scientific™

Product Code. 11956651 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11956651 250 mg Glass bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11956651 Supplier Thermo Scientific Acros Supplier No. 434102500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Specifications

CAS 60261-46-3
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Tan
Density 1.0400g/mL
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C17H20NP
MDL Number MFCD16621443
Quantity 250 mg
Specific Gravity 1.04
Synonym s-2-diphenylphosphino methyl pyrrolidine, 2s-2-diphenylphosphanyl methyl pyrrolidine, pyrrolidine, 2-diphenylphosphino methyl-, 2s, aminophosphane
InChI Key IWRBGJKCDORZHK-GGYSOQFKNA-N
SMILES C([C@@H]1CCCN1)P(C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name diphenyl-[[(2S)-pyrrolidin-2-yl]methyl]phosphane
Molecular Weight (g/mol) 269.33
PubChem CID 11272693
Formula Weight 269.32
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (S)-2-((Diphenylphosphino)methyl)pyrrolidine
Show More Show Less
compliance-icons
Product Identifier
  • (S)-2-((Diphenylphosphino)methyl)pyrrolidine
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.