missing translation for 'onlineSavingsMsg'
Learn More

S-(2-Guanidino-4-thiazolyl)methylisothiourea Dihydrochloride, TRC

Product Code. 30403916
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30403916 10 g
30395792 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30403916 Supplier Toronto Research Chemicals Supplier No. TRCG83580010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88046-01-9
Molecular Formula C6 H10 N6 S2 . 2 Cl H
Molecular Weight (g/mol) 303.236
Synonym Carbamimidothioic acid, [2-[(aminoiminomethyl)amino]-4-thiazolyl]methyl ester, dihydrochloride (9CI), [2-[(Diaminomethylidene)amino]thiazol-4-yl]methyl Carbamimidothioate Hydrochloride, S-(2-Guanidino-4-thiazolyl)methylisothiourea Dihydrochloride, Famotidine Imp. H (EP) as Hydrochloride
IUPAC Name (2-carbamimidamido-1,3-thiazol-4-yl)methyl carbamimidothioate;dihydrochloride
SMILES Cl.Cl.NC(=N)Nc1nc(CSC(=N)N)cs1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C6H10N6S2.2ClH/c7-4(8)12-6-11-3(2-14-6)1-13-5(9)10;;/h2H,1H2,(H3,9,10)(H4,7,8,11,12);2*1H
Formula Weight 301.994
Chemical Name or Material (2-[(Diaminomethylidene)amino]thiazol-4-yl)methyl Carbamimidothioate Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.