missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-(Methoxymethyl)pyrrolidine, 98%

Product Code. 11382157
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382157 5 g 5g
11392157 250 mg 250mg
11372157 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11382157 Supplier Thermo Scientific Alfa Aesar Supplier No. L09608.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 63126-47-6 | C6H13NO | 115.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63126-47-6
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00010408
InChI Key CHPRFKYDQRKRRK-LURJTMIESA-N
Synonym s-2-methoxymethyl pyrrolidine, s-+-2-methoxymethyl pyrrolidine, 2s-2-methoxymethyl pyrrolidine, o-methyl-l-prolinol, s-2-methoxymethyl-pyrrolidine, s-2-methoxymethylpyrrolidine, s-+-2-methoxymethyl pyrolidine, pyrrolidine, 2-methoxymethyl-, 2s, s-+-2-methoxymethyl pyrrplidine, 2s pyrrolidin-2-yl methoxymethane
PubChem CID 671217
IUPAC Name (2S)-2-(methoxymethyl)pyrrolidine
SMILES COCC1CCCN1

Specifications

Density 0.933
Boiling Point 61°C to 62°C (40 mmHg)
Flash Point 45°C (113°F)
Refractive Index 1.4457
Quantity 5 g
UN Number UN1993
Beilstein 3587324
Sensitivity Air Sensitive
Optical Rotation +2.2° (c=2 in Benzene)
Formula Weight 115.18
Percent Purity 98%
Chemical Name or Material (S)-(+)-2-(Methoxymethyl)pyrrolidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.