missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-(Methoxymethyl)pyrrolidine, 99%, Thermo Scientific™

Product Code. 10464575 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Glass bottle; 2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10464575 Glass bottle; 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10464575 Supplier Thermo Scientific Acros Supplier No. 309910025
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 63126-47-6
CAS Min % 98.5
CAS Max % 100.0
Density 0.9300g/mL
Boiling Point 62.0°C (40.0 mmHg)
Flash Point 45°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C6H13NO
Refractive Index 1.4450 to 1.4470
MDL Number MFCD00010408
Quantity 2.5g
Specific Gravity 0.93
Specific Rotation Condition + 36.60 (20.00°C c=2,2N HCl)
Specific Rotation + 36.60
Synonym s-2-methoxymethyl pyrrolidine, s-+-2-methoxymethyl pyrrolidine, 2s-2-methoxymethyl pyrrolidine, o-methyl-l-prolinol, s-2-methoxymethyl-pyrrolidine, s-2-methoxymethylpyrrolidine, s-+-2-methoxymethyl pyrolidine, pyrrolidine, 2-methoxymethyl-, 2s, s-+-2-methoxymethyl pyrrplidine, 2s pyrrolidin-2-yl methoxymethane
InChI Key CHPRFKYDQRKRRK-LURJTMIESA-N
SMILES COCC1CCCN1
IUPAC Name (2S)-2-(methoxymethyl)pyrrolidine
Molecular Weight (g/mol) 115.18
PubChem CID 671217
Formula Weight 115.18
Percent Purity 99%
Chemical Name or Material (S)-(+)-2-(Methoxymethyl)pyrrolidine, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.