missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Octyl isocyanate, 95%

Product Code. 15479149
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15479149 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15479149 Supplier Thermo Scientific Alfa Aesar Supplier No. L20245.03
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 745783-87-3 | C9H17NO | 155.241 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 745783-87-3
Density 0.826
Boiling Point 68°C (5 mmHg)
Molecular Formula C9H17NO
Refractive Index 1.423
MDL Number MFCD05664089
Quantity 1 g
UN Number UN2206
Sensitivity Moisture sensitive; May form a precipitate
Synonym s-+-2-octyl isocyanate, 2s-2-isocyanatooctane
InChI Key FTWPDIQXUDUDPL-VIFPVBQESA-N
SMILES CCCCCCC(C)N=C=O
IUPAC Name (2S)-2-isocyanatooctane
Molecular Weight (g/mol) 155.241
PubChem CID 7018274
Formula Weight 155.24
Percent Purity 95%
Chemical Name or Material (S)-(+)-2-Octyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.