missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Phenylglycinol, 98%

Product Code. 13428508 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13428508 25 g Glass bottle 25g
10287750 1 g Glass bottle 1g
10118093 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 13428508 Supplier Thermo Scientific Acros Supplier No. 307570250
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 20989-17-7 | C8H11NO | 137.182 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20989-17-7
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
MDL Number MFCD00064404
InChI Key IJXJGQCXFSSHNL-MRVPVSSYSA-N
Synonym s-+-2-phenylglycinol, s-2-phenylglycinol, s-2-amino-2-phenylethanol, s-phenylglycinol, h-phenylglycinol, 2s-2-amino-2-phenylethanol, 2s-2-amino-2-phenylethan-1-ol, l-2-phenylglycinol, h-phg-ol
PubChem CID 134797
IUPAC Name (2S)-2-amino-2-phenylethanol
SMILES C1=CC=C(C=C1)C(CO)N

Specifications

Infrared Spectrum Authentic
Color White to Yellow
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Specific Rotation + 25.50
Melting Point 75.0°C to 79.0°C
Quantity 25 g
Physical Form Crystalline Powder
Assay Percent Range 98%
Linear Formula C6H5CH(NH2)CH2OH
Formula Weight 137.18
Percent Purity 98%
Chemical Name or Material (S)-(+)-2-Phenylglycinol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.