missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-((S)-4-Methyl-2-((S)-2-(2-morpholinoacetamido)-4-phenylbutanamido)pentanamido)-3-phenylpropanoic Acid, TRC

Product Code. 30469552
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30469552 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30469552 Supplier Toronto Research Chemicals Supplier No. TRCM320448500MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-2-((S)-4-Methyl-2-((S)-2-(2-morpholinoacetamido)-4-phenylbutanamido)pentanamido)-3-phenylpropanoic Acid
CAS 868540-16-3
Molecular Formula C31 H42 N4 O6
Quantity 500 mg
Synonym (αS)-α-[[2-(4-Morpholinyl)acetyl]amino]benzenebutanoyl-L-leucyl-L-phenylalanine, (αS)-α-[(4-Morpholinylacetyl)amino]benzenebutanoyl-L-leucyl-L-phenylalanine
InChI Formula InChI=1S/C31H42N4O6/c1-22(2)19-26(30(38)34-27(31(39)40)20-24-11-7-4-8-12-24)33-29(37)25(14-13-23-9-5-3-6-10-23)32-28(36)21-35-15-17-41-18-16-35/h3-12,22,25-27H,13-21H2,1-2H3,(H,32,36)(H,33,37)(H,34,38)(H,39,40)/t25-,26-,27-/m0/s1
SMILES CC(C[C@H](NC([C@@H](NC(CN1CCOCC1)=O)CCC2=CC=CC=C2)=O)C(N[C@H](C(O)=O)CC3=CC=CC=C3)=O)C
IUPAC Name (2S)-2-[[(2S)-4-methyl-2-[[(2S)-2-[(2-morpholin-4-ylacetyl)amino]-4-phenylbutanoyl]amino]pentanoyl]amino]-3-phenylpropanoic acid
Molecular Weight (g/mol) 566.69
Formula Weight 566.3104
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.