missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-[(tert-Butoxycarbonyl)amino]-5-(dimethylamino)pentanoic acid, Thermo Scientific™

Product Code. 10673425
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10673425 1 g Amber Glass Bottle 1g
10295894 250 mg Amber Glass Bottle 250mg
10430934 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10673425 Supplier Thermo Scientific Maybridge Supplier No. MO08534DA
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 65671-54-7
Molecular Formula C12H24N2O4
Molecular Weight (g/mol) 260.334
InChI Key HBCLYVIARHJTTL-VIFPVBQESA-N
Synonym s-2-tert-butoxycarbonyl amino-5-dimethylamino pentanoic acid, 2s-2-tert-butoxycarbonyl amino-5-dimethylamino pentanoic acid, s-2-boc-amino-5-dimethylamino pentanoic acid, s-2-tert-butoxycarbonyl amino-5-dimethylami, n∼2∼-tert-butoxycarbonyl-n∼5∼,n∼5∼-dimethyl-l-ornithine, s-2-tert-butoxycarbonylamino-5-dimethylamino pentanoic acid, l-ornithine, n2-1,1-dimethylethoxy carbonyl-n5,n5-dimethyl, 2s-5-dimethylamino-2-tert-butoxy carbonylamino pentanoic acid
PubChem CID 16105709
IUPAC Name (2S)-5-(dimethylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
SMILES CC(C)(C)OC(=O)NC(CCCN(C)C)C(=O)O

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 260.33
Percent Purity 97%
Chemical Name or Material (S)-2-[(tert-Butoxycarbonyl)amino]-5-(dimethylamino)pentanoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.