missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-3-Amino-1-BOC-piperidine, 95%

Product Code. 11966381 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11966381 5 g Glass bottle 5g
11956381 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11966381 Supplier Thermo Scientific Acros Supplier No. 437490050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 625471-18-3 | C10H20N2O2 | 200.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625471-18-3
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.28
MDL Number MFCD03094718
InChI Key AKQXKEBCONUWCL-UHFFFAOYNA-N
Synonym s-1-boc-3-aminopiperidine, s-3-amino-1-n-boc-piperidine, tert-butyl 3s-3-aminopiperidine-1-carboxylate, s-tert-butyl 3-aminopiperidine-1-carboxylate, s-3-amino-1-boc-piperidine, s-+-3-amino-1-boc-piperidine, 3s-3-aminopiperidine, n1-boc protected, s-1-boc-3-amniopiperidine, s-3-amino-piperidine-1-carboxylic acid tert-butyl ester, s-3-amino-n-tboc-piperidine
PubChem CID 1501975
IUPAC Name tert-butyl (3S)-3-aminopiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(N)C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Rotation 30
Formula Weight 200.28
Percent Purity 95%
Physical Form Gum or Semi-solid
Chemical Name or Material (S)-(+)-3-Amino-1-BOC-piperidine
Show More Show Less
compliance-icons
Product Identifier
  • (S)-(+)-3-Amino-1-BOC-piperidine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.