missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-3-Amino-1-Boc-piperidine, 97%

Product Code. 15413957
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15413957 5 g 5g
15423957 250 mg 250mg
15403957 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15413957 Supplier Thermo Scientific Alfa Aesar Supplier No. H30538.06
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 625471-18-3 | C10H20N2O2 | 200.28 g/mol

(S)-(+)-3-Amino-1-Boc-piperidine is used to prepare selective noncovalent inhibitors of the bacterial cysteine protease IdeS. The product obtained by the reductive elimination reaction with ethylglyoxalte is used as a reference compound, which is used to determine the absolute configuration of the two enantiomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-3-Amino-1-Boc-piperidine is used to prepare selective noncovalent inhibitors of the bacterial cysteine protease IdeS. The product obtained by the reductive elimination reaction with ethylglyoxalte is used as a reference compound, which is used to determine the absolute configuration of the two enantiomers.

Solubility
Soluble in dimethylsulfoxide.

Notes
Air and carbon dioxide sensitive. Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625471-18-3
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.28
MDL Number MFCD03094718
InChI Key AKQXKEBCONUWCL-UHFFFAOYNA-N
Synonym s-1-boc-3-aminopiperidine, s-3-amino-1-n-boc-piperidine, tert-butyl 3s-3-aminopiperidine-1-carboxylate, s-tert-butyl 3-aminopiperidine-1-carboxylate, s-3-amino-1-boc-piperidine, s-+-3-amino-1-boc-piperidine, 3s-3-aminopiperidine, n1-boc protected, s-1-boc-3-amniopiperidine, s-3-amino-piperidine-1-carboxylic acid tert-butyl ester, s-3-amino-n-tboc-piperidine
PubChem CID 1501975
IUPAC Name tert-butyl (3S)-3-aminopiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(N)C1

Specifications

Quantity 5 g
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Soluble in dimethylsulfoxide.
Formula Weight 200.28
Percent Purity 97%
Chemical Name or Material (S)-(+)-3-Amino-1-Boc-piperidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.