missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-Amino-3-phenylpropan-1-ol, 95%, 98% ee, Thermo Scientific Chemicals

Product Code. 10304692 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
Product Code. Quantity Packaging unitSize
10304692 250 mg Glass bottle 250mg
10552232 1 g Glass bottle 1g
2 options
This item is not returnable. View return policy

Product Code. 10304692

Brand: Thermo Scientific Acros 362172500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82769-76-4
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD01311768
InChI Key SEQXIQNPMQTBGN-VIFPVBQESA-N
Synonym s-3-amino-3-phenylpropan-1-ol, 3s-3-amino-3-phenylpropan-1-ol, s-1-phenyl-3-propanolamine, s-beta-phenylalaninol, s-3-phenyl-beta-alaninol, s-3-amino-3-phenyl-1-propanol, s-3-amino-3-phenyl-propan-1-ol, 3s-3-amino-3-phenyl-propan-1-ol, pubchem13854, ksc491q5n
PubChem CID 2734520
IUPAC Name (3S)-3-amino-3-phenylpropan-1-ol
SMILES C1=CC=C(C=C1)C(CCO)N

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless or White
Enantiomeric Excess 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 250 mg
Formula Weight 151.21
Percent Purity 95%,98% ee
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material (S)-3-Amino-3-phenylpropan-1-ol, 95%, (98% e.e.)
Show More Show Less
compliance-icons
Product Identifier
  • (S)-3-Amino-3-phenylpropan-1-ol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.