missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-Amino-5-hexynoic acid hydrochloride, 95%

Product Code. 15479495
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15479495 250 mg 250mg
15458497 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15479495 Supplier Thermo Scientific Alfa Aesar Supplier No. H52010.MD
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 270596-46-8 | C6H10ClNO2 | 163.601 g/mol

(S)-3-Amino-5-hexynoic acid hydrochloride is used for labeling and detection purpose.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-3-Amino-5-hexynoic acid hydrochloride is used for labeling and detection purpose.

Solubility
Soluble in water (90 g/L at 25°C).

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 270596-46-8
Molecular Formula C6H10ClNO2
Molecular Weight (g/mol) 163.601
MDL Number MFCD01861006
InChI Key FVKOZZHCHSRKJA-JEDNCBNOSA-N
Synonym s-3-amino-5-hexynoic acid hydrochloride, s-3-aminohex-5-ynoic acid hydrochloride, 3s-3-aminohex-5-ynoic acid hydrochloride, s-3-amino-5-hexynoic acid-hcl, l-beta-homopropargylglycine hydrochloride, 5-hexynoic acid, 3-amino-, 3s-, hydrochloride 1:1, s-3-amino-5-hexynoic acidhcl, l-ss-homopropargylglycine hydrochloride
PubChem CID 2761700
IUPAC Name (3S)-3-aminohex-5-ynoic acid;hydrochloride
SMILES C#CCC(CC(=O)O)N.Cl

Specifications

Solubility Information Soluble in water (90g/L at 25°C).
Quantity 250 mg
Formula Weight 163.6
Sensitivity Air Sensitive
Percent Purity 95%
Chemical Name or Material (S)-3-Amino-5-hexynoic acid hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.