missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-Aminomethyl-1-Boc-piperidine, 97%

Product Code. 15499587
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15499587 250 mg 250mg
15479587 1 g 1g
15489587 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15499587 Supplier Thermo Scientific Alfa Aesar Supplier No. H52822.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 140645-24-5 | C11H22N2O2 | 214.309 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140645-24-5
Molecular Formula C11H22N2O2
Molecular Weight (g/mol) 214.309
MDL Number MFCD03839877
InChI Key WPWXYQIMXTUMJB-VIFPVBQESA-N
Synonym s-1-boc-3-aminomethyl piperidine, s-n-boc-3-aminomethylpiperidine, s-3-aminomethyl-1-boc-piperidine, s-tert-butyl 3-aminomethyl piperidine-1-carboxylate, s-1-n-boc-3-aminomethyl piperidine, s-3-aminomethyl-1-n-boc-piperidine, tert-butyl 3s-3-aminomethyl piperidine-1-carboxylate, s-3-aminomethyl-piperidine-1-carboxylic acid tert-butyl ester, s-1-n-boc-piperidine-3-methylamine, 3s-3-aminomethyl-n-tert-butoxycarbonylpiperidine
PubChem CID 1502022
IUPAC Name tert-butyl (3S)-3-(aminomethyl)piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(C1)CN

Specifications

Quantity 250 mg
Formula Weight 214.31
Percent Purity 97%
Chemical Name or Material (S)-3-Aminomethyl-1-Boc-piperidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.