missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-3-(Boc-amino)-5-methylhexanoic acid, 95%
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Description
(S)-3-(Boc-amino)-5-methylhexanoic acid is used as building blocks for the synthesis of carba peptides and building block for β-peptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(S)-3-(Boc-amino)-5-methylhexanoic acid is used as building blocks for the synthesis of ″carba peptides″ and building block for β-peptides.
Solubility
Very slightly soluble (1.1 g/L at 25°C).
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
(S)-3-(Boc-amino)-5-methylhexanoic acid is used as building blocks for the synthesis of ″carba peptides″ and building block for β-peptides.
Solubility
Very slightly soluble (1.1 g/L at 25°C).
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications
Specifications
| Melting Point | 53°C to 55°C |
| Quantity | 250 mg |
| Beilstein | 4251252 |
| Solubility Information | Very slightly soluble (1.1g/L at 25°C). |
| Formula Weight | 245.32 |
| Percent Purity | 95% |
| Chemical Name or Material | (S)-3-(Boc-amino)-5-methylhexanoic acid |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction