missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-(Boc-amino)-5-phenylpentanoic acid, 95%, Thermo Scientific Chemicals

Product Code. 15448737
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15448737

Brand: Thermo Scientific Alfa Aesar H52180.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 218608-84-5
Molecular Formula C16H23NO4
Molecular Weight (g/mol) 293.36
MDL Number MFCD01076264,MFCD05863635,MFCD01076262
InChI Key MYWZFJXOLAXENE-UHFFFAOYNA-N
Synonym r-3-tert-butoxycarbonyl amino-5-phenylpentanoic acid, r-3-boc-amino-5-phenylpentanoic acid, 3r-3-tert-butoxycarbonyl amino-5-phenylpentanoic acid, boc-d-?-nva 5-phenyl-oh, n-boc-3-r-amino-5-phenylpentanoic acid, n-tert-butoxycarbonyl-3 r-amino-5-phenylpentanoic acid, 3r-3-tert-butoxycarbonylamino-5-phenyl-pentanoic acid, r-3-tert-butoxycarbonyl amino-5-phenylpentanoicacid, r-3-boc-amino-5-phenylpentanoic acid hplc, 3r-3-2-methylpropan-2-yl oxycarbonylamino-5-phenylpentanoic acid
PubChem CID 7021558
IUPAC Name (3R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-5-phenylpentanoic acid
SMILES CC(C)(C)OC(=O)NC(CCC1=CC=CC=C1)CC(O)=O

Specifications

Quantity 1 g
Formula Weight 293.36
Percent Purity 95%
Chemical Name or Material (S)-3-(Boc-amino)-5-phenylpentanoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.