missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-3-(Boc-amino)pyrrolidine, 99%, ee 99%

Product Code. 11325525
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11325525 1 g 1g
11335525 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11325525 Supplier Thermo Scientific Alfa Aesar Supplier No. L19693.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 122536-76-9 | C9H18N2O2 | 186.255 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122536-76-9
Molecular Formula C9H18N2O2
Molecular Weight (g/mol) 186.255
MDL Number MFCD00143194
InChI Key DQQJBEAXSOOCPG-ZETCQYMHSA-N
Synonym s-3-boc-amino pyrrolidine, s-tert-butyl pyrrolidin-3-ylcarbamate, s---3-boc-amino pyrrolidine, s-3-n-boc-aminopyrrolidine, tert-butyl n-3s-pyrrolidin-3-yl carbamate, s-+-3-boc-amino pyrrolidine, 3s---3-tert-butoxycarbonylamino pyrrolidine, tert-butyl s-pyrrolidin-3-ylcarbamate, s-3bocap, 3s---3-boc-amino pyrrolidine
PubChem CID 1514396
IUPAC Name tert-butyl N-[(3S)-pyrrolidin-3-yl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCNC1

Specifications

Melting Point 50°C to 54°C
UN Number UN3259
Beilstein 5377813
Quantity 1 g
Formula Weight 186.25
Optical Rotation −23° (c=1 in Methanol)
Sensitivity Air sensitive
Percent Purity 99%
Assay ee 99%
Chemical Name or Material (S)-(-)-3-(Boc-amino)pyrrolidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.