missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-3-(BOC-Amino)pyrrolidine, 99%

Product Code. 10795622 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10795622 5 g 5g
10139439 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10795622 Supplier Thermo Scientific Acros Supplier No. 437900050
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 122536-76-9 | C9H18N2O2 | 186.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122536-76-9
Molecular Formula C9H18N2O2
Molecular Weight (g/mol) 186.26
InChI Key DQQJBEAXSOOCPG-ZETCQYMHSA-N
Synonym s-3-boc-amino pyrrolidine, s-tert-butyl pyrrolidin-3-ylcarbamate, s---3-boc-amino pyrrolidine, s-3-n-boc-aminopyrrolidine, tert-butyl n-3s-pyrrolidin-3-yl carbamate, s-+-3-boc-amino pyrrolidine, 3s---3-tert-butoxycarbonylamino pyrrolidine, tert-butyl s-pyrrolidin-3-ylcarbamate, s-3bocap, 3s---3-boc-amino pyrrolidine
PubChem CID 1514396
IUPAC Name tert-butyl N-[(3S)-pyrrolidin-3-yl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCNC1

Specifications

Melting Point 77.8°C to 80.7°C
CAS Min % 98.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Specific Rotation -20
Quantity 5 g
Formula Weight 186.26
Percent Purity 99%
Physical Form Powder
Chemical Name or Material (S)-(-)-3-(BOC-amino)pyrrolidine
Show More Show Less
compliance-icons
Product Identifier
  • (S)-(-)-3-(BOC-Amino)pyrrolidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.