missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-Boc-thiazolidine-2-carboxylic acid, 97%

Product Code. 15419547
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15419547 5 g 5g
15429547 250 mg 250mg
15409547 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15419547 Supplier Thermo Scientific Alfa Aesar Supplier No. H52793.06
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 891192-95-3 | C9H15NO4S | 233.282 g/mol

(S)-3-Boc-thiazolidine-2-carboxylic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-3-Boc-thiazolidine-2-carboxylic acid is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 891192-95-3
Molecular Formula C9H15NO4S
Molecular Weight (g/mol) 233.282
MDL Number MFCD02682342
InChI Key HYAXPNDMEODKHI-LURJTMIESA-N
Synonym s-3-boc-2-thiazolidinecarboxylic acid, s-3-tert-butoxycarbonyl thiazolidine-2-carboxylic acid, 2s-3-tert-butoxycarbonyl-1,3-thiazolidine-2-carboxylic acid, 2,3-thiazolidinedicarboxylic acid, 3-1,1-dimethylethyl ester, 2s, s-3-boc-thiazolidine-2-carboxylic acid, n-boc s thiazolidine-2-carboxylic acid, n-boc-s-thiazolidine-2-carboxylic acid, s-thiazolidine-2,3-dicarboxylic acid 3-tert-butyl ester, 2s-3-tert-butoxy carbonyl-1,3-thiazolidine-2-carboxylic acid, 2s-2,3-thiazolidinedicarboxylic acid, 3-1,1-dimethylethyl ester
PubChem CID 14775612
IUPAC Name (2S)-3-[(2-methylpropan-2-yl)oxycarbonyl]-1,3-thiazolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCSC1C(=O)O

Specifications

Melting Point 90°C to 93°C
Quantity 5 g
Optical Rotation −47° (c=5 in Methanol)
Formula Weight 233.28
Percent Purity 97%
Chemical Name or Material (S)-3-Boc-thiazolidine-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.