missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-3-Chloro-1-phenyl-1-propanol, 98+%

Product Code. 15451018
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15451018 250 mg 250mg
15431018 1 g 1g
15441018 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15451018 Supplier Thermo Scientific Alfa Aesar Supplier No. H54847.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 100306-34-1 | C9H11ClO | 170.636 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100306-34-1
Molecular Formula C9H11ClO
Molecular Weight (g/mol) 170.636
MDL Number MFCD00013309
InChI Key JZFUHAGLMZWKTF-VIFPVBQESA-N
Synonym s---3-chloro-1-phenyl-1-propanol, s-3-chloro-1-phenylpropan-1-ol, s-3-chloro-1-phenyl-1-propanol, 1s-3-chloro-1-phenylpropan-1-ol, 1s-3-chloro-1-phenyl-propan-1-ol, s-3-chloro-1-phenylpropanol, s-3-chloro-1-phenyl-propan-1-ol, pubchem6275
PubChem CID 2777894
IUPAC Name (1S)-3-chloro-1-phenylpropan-1-ol
SMILES C1=CC=C(C=C1)C(CCCl)O

Specifications

Melting Point 58°C to 60°C
Quantity 250 mg
Optical Rotation −26° (c=1 in Chloroform)
Formula Weight 170.64
Percent Purity ≥98%
Chemical Name or Material (S)-(-)-3-Chloro-1-phenyl-1-propanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.