missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-Hydroxyisobutyric Acid Potassium Salt, TRC

Product Code. 30404838
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30404838 500 mg 500mg
30395899 100 mg -
30393876 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30404838 Supplier Toronto Research Chemicals Supplier No. TRCH943225500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1402163-95-4
Molecular Formula C4H7KO3
Molecular Weight (g/mol) 142.19
Synonym 3-Hydroxy-2-methylpropanoic Acid Potassium Salt
IUPAC Name potassium;(2S)-3-hydroxy-2-methylpropanoate
SMILES [K+].C[C@@H](CO)C(=O)[O-]

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C4H8O3.K/c1-3(2-5)4(6)7;/h3,5H,2H2,1H3,(H,6,7);/q;+1/p-1/t3-;/m0./s1
Formula Weight 142.0032
Chemical Name or Material (S)-3-Hydroxyisobutyric Acid Potassium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.