missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-Hydroxypyrrolidine hydrochloride, 97%

Product Code. 15469577
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15469577 1 g 1g
15479577 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15469577 Supplier Thermo Scientific Alfa Aesar Supplier No. H52815.03
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 122536-94-1 | C4H10ClNO | 123.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122536-94-1
Molecular Formula C4H10ClNO
Molecular Weight (g/mol) 123.58
MDL Number MFCD00272298
InChI Key QPMSJEFZULFYTB-WCCKRBBISA-N
Synonym s-3-hydroxypyrrolidine hydrochloride, s-pyrrolidin-3-ol hydrochloride, 3s-pyrrolidin-3-ol hydrochloride, s-+-3-pyrrolidinol hydrochloride, s-3-hydroxypyrrolidine-hcl, s-3-pyrrolidinol hydrochloride, 3s-pyrrolidinol hydrochloride, s-3-pyrrolidinol hcl, s-pyrrolidin-3-ol, hcl, s---3-pyrrolidinol hydrochloride
PubChem CID 22309122
IUPAC Name (3S)-pyrrolidin-3-ol;hydrochloride
SMILES C1CNCC1O.Cl

Specifications

Melting Point 106°C to 107°C
Boiling Point 108°C to 110°C (8 mmHg)
Quantity 1 g
Formula Weight 123.58
Percent Purity 97%
Chemical Name or Material (S)-3-Hydroxypyrrolidine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.